siRNA Details
| Gene Symbol: SCCPDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016002.2 | NP_057086.2 | 1, 2 | 546 | ||
| GenBank mRNA | AF151807.1 | - | 1, 2 | 196 | ||
| AK075178.1 | - | 1, 2 | 302 | |||
| BC026185.2 | - | 1, 2 | 207 | 4798194 | ||
| BG258610.1 | - | 1, 2 | 546 | |||

SCCPDH
Genomic Map