siRNA Details
Gene Symbol: SCCPDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016002.2 | NP_057086.2 | 5 | 897 | ||
GenBank mRNA | AF151807.1 | - | 5 | 547 | ||
AK075178.1 | - | 5 | 653 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SCCPDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016002.2 | NP_057086.2 | 5 | 897 | ||
GenBank mRNA | AF151807.1 | - | 5 | 547 | ||
AK075178.1 | - | 5 | 653 |