siRNA Details
| Gene Symbol: NDUFAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016013.3 | NP_057097.2 | 2 | 640 | ||
| NR_045620.1 | 2 | 640 | ||||
| XM_006720555.2 | XP_006720618.1 | 2 | 570 | |||
| XM_011521658.1 | XP_011519960.1 | 2 | 504 | |||
| XM_011521659.2 | XP_011519961.1 | 2 | 667 | |||
| XM_017022293.1 | XP_016877782.1 | 2 | 571 | |||
| GenBank mRNA | AF151823.1 | - | 2 | 603 | ||
| AK309889.1 | - | 2 | 640 | |||
| AK310983.1 | - | 2 | 640 | |||
| BC000780.2 | - | 2 | 624 | 3504355 | ||

NDUFAF1
Genomic Map