siRNA Details
Gene Symbol: TRAPPC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321102.1 | NP_001308031.1 | 9 | 1870 | ||
NM_016030.5 | NP_057114.5 | 9 | 1907 | |||
XM_011510350.2 | XP_011508652.1 | 8 | 2223 | |||
XM_011510355.2 | XP_011508657.1 | 8 | 848 | |||
XM_011510356.1 | XP_011508658.1 | 8 | 740 | |||
XM_017004257.1 | XP_016859746.1 | 9 | 2337 | |||
GenBank mRNA | AF151845.1 | - | 7 | 636 | ||
AK094181.1 | - | 10 | 1875 | |||
AK094242.1 | - | 8 | 1281 | |||
AK098327.1 | - | 9 | 1872 | |||
AK125105.1 | - | 4 | 2787 | |||
BC017475.1 | - | 8 | 905 | |||
BQ954367.1 | - | 4 | 219 | |||
BU782660.1 | - | 4 | 630 |
