siRNA Details
| Gene Symbol: UTP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016037.3 | NP_057121.2 | 7 | 695 | ||
| XM_011541555.2 | XP_011539857.1 | 6 | 629 | |||
| GenBank mRNA | AF067802.1 | - | 7 | 658 | ||
| AF151852.1 | - | 7 | 673 | |||
| AK023587.1 | - | 5 | 1564 | |||
| AK291900.1 | - | 7 | 625 | |||
| AK296018.1 | - | 6 | 497 | |||
| AK308377.1 | - | 6 | 630 | |||
| BC005182.1 | - | 7 | 640 | 3995984 | ||
| CX163866.1 | - | 2 | 61 | |||

UTP11
Genomic Map