siRNA Details
| Gene Symbol: IER3IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016097.4 | NP_057181.1 | 2, 3 | 288 | ||
| GenBank mRNA | AF119875.1 | - | 1, 2 | 1310 | ||
| AF125100.1 | - | 2, 3 | 271 | |||
| AF164798.1 | - | 2, 3 | 257 | |||
| AF371963.1 | - | 2, 3 | 261 | |||
| AK027108.1 | - | 2, 3 | 278 | |||
| AL136667.1 | - | 2, 3 | 268 | |||
| BC010888.1 | - | 2, 3 | 254 | 4285974 | ||
| BC017391.1 | - | 2, 3 | 221 | 4133818 | ||
| BM829309.1 | - | 2, 3 | 287 | |||

IER3IP1
Genomic Map