siRNA Details
| Gene Symbol: A4GNT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016161.2 | NP_057245.1 | 3 | 883 | ||
| XM_017006543.1 | XP_016862032.1 | 3 | 1434 | |||
| XM_017006544.1 | XP_016862033.1 | 3 | 1004 | |||
| GenBank mRNA | AF141315.1 | - | 3 | 861 | ||
| AK225668.1 | - | 3 | 883 | |||
| BC119639.1 | - | 3 | 778 | 40115688 | ||
| BC119640.2 | - | 3 | 778 | 40115689 | ||

A4GNT
Genomic Map