siRNA Details
| Gene Symbol: MYO15A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016239.3 | NP_057323.3 | 54, 55 | 9560 | ||
| XM_017024714.1 | XP_016880203.1 | 51, 52 | 9860 | |||
| XM_017024715.1 | XP_016880204.1 | 52, 53 | 9923 | |||
| GenBank mRNA | AF144094.1 | - | 54, 55 | 9560 | ||
| AK294036.1 | - | 12, 13 | 1243 | |||
| AK297179.1 | - | 5, 6 | 487 | |||

MYO15A
Genomic Map