siRNA Details
| Gene Symbol: GPN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164372.1 | NP_001157844.1 | 2 | 330 | ||
| NM_001164373.1 | NP_001157845.1 | 2 | 239 | |||
| NM_016301.3 | NP_057385.3 | 2 | 209 | |||
| XM_005253896.4 | XP_005253953.3 | 3 | 664 | |||
| XM_017019394.1 | XP_016874883.1 | 2 | 144 | |||
| GenBank mRNA | AF117229.1 | - | 2 | 159 | ||
| AK223209.1 | - | 2 | 181 | |||
| AK314946.1 | - | 2 | 182 | |||
| AM992619.1 | - | 2 | 128 | |||
| AM992620.1 | - | 2 | 128 | |||
| AY359078.1 | - | 2 | 210 | |||
| AY364262.1 | - | 2 | 128 | |||
| BC008416.1 | - | 2 | 184 | 4048678 | ||
| BC031024.1 | - | 2 | 185 | 4695834 | ||
| BI546381.1 | - | 2 | 333 | |||
| BX537973.1 | - | 2 | 220 | |||
| DB455987.1 | - | 2 | 209 | |||

GPN3
Genomic Map