siRNA Details
| Gene Symbol: HERC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016323.3 | NP_057407.2 | 4, 5 | 838 | ||
| XM_011532022.2 | XP_011530324.2 | 4, 5 | 1351 | |||
| GenBank mRNA | AB027289.1 | - | 4, 5 | 798 | ||
| AY337518.1 | - | 4, 5 | 838 | |||
| BC140716.1 | - | 4, 5 | 731 | 9021584 | ||

HERC5
Genomic Map