siRNA Details
| Gene Symbol: COPZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016429.3 | NP_057513.1 | 4 | 328 | ||
| XM_011524874.2 | XP_011523176.1 | 4 | 327 | |||
| GenBank mRNA | AB037938.1 | - | 4 | 312 | ||
| AB047849.1 | - | 4 | 310 | |||
| BC015924.1 | - | 4 | 304 | 4850757 | ||
| BQ576336.1 | - | 5 | 530 | |||
| HY179834.1 | - | 4 | 323 | |||

COPZ2
Genomic Map