siRNA Details
| Gene Symbol: TFDP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016521.2 | NP_057605.3 | 1 | 1163 | ||
| GenBank mRNA | AF219119.2 | - | 1 | 1163 | ||
| BC074762.2 | - | 1 | 922 | |||
| BC117336.1 | - | 1 | 959 | |||
| BC126361.1 | - | 1 | 959 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TFDP3| Gene Symbol: TFDP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016521.2 | NP_057605.3 | 1 | 1163 | ||
| GenBank mRNA | AF219119.2 | - | 1 | 1163 | ||
| BC074762.2 | - | 1 | 922 | |||
| BC117336.1 | - | 1 | 959 | |||
| BC126361.1 | - | 1 | 959 | |||