siRNA Details
| Gene Symbol: DNAJC27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198559.1 | NP_001185488.1 | 5 | 655 | ||
| NM_016544.2 | NP_057628.1 | 5 | 655 | |||
| XM_011532902.1 | XP_011531204.1 | 5 | 630 | |||
| XM_017004269.1 | XP_016859758.1 | 5 | 686 | |||
| XM_017004270.1 | XP_016859759.1 | 5 | 546 | |||
| GenBank mRNA | AF178983.1 | - | 5 | 489 | ||
| AK294726.1 | - | 5 | 439 | |||
| AL137731.1 | - | 6 | 678 | |||
| AY094594.1 | - | 5 | 465 | |||
| BC034049.1 | - | 5 | 516 | 4815297 | ||

DNAJC27
Genomic Map