siRNA Details
| Gene Symbol: DNAJC27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198559.1 | NP_001185488.1 | 6 | 792 | ||
| NM_016544.2 | NP_057628.1 | 7 | 953 | |||
| XM_017004269.1 | XP_016859758.1 | 7 | 984 | |||
| XM_017004270.1 | XP_016859759.1 | 7 | 844 | |||
| GenBank mRNA | AF178983.1 | - | 7 | 787 | ||
| AK294726.1 | - | 7 | 737 | |||
| AL137731.1 | - | 9 | 1062 | |||
| AY094594.1 | - | 6 | 602 | |||
| BC034049.1 | - | 7 | 814 | 4815297 | ||

DNAJC27
Genomic Map