siRNA Details
| Gene Symbol: BFAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016561.2 | NP_057645.1 | 6, 7 | 1232 | ||
| XM_005255350.2 | XP_005255407.1 | 5, 6 | 1007 | |||
| XM_011522520.1 | XP_011520822.1 | 4, 5 | 837 | |||
| GenBank mRNA | AF173003.1 | - | 6, 7 | 1104 | ||
| AK291114.1 | - | 6, 7 | 1040 | |||
| AK297067.1 | - | 6, 7 | 793 | |||
| AK300778.1 | - | 4, 5 | 805 | |||
| AK313890.1 | - | 5, 6 | 1040 | |||
| BC003054.1 | - | 6, 7 | 1063 | 3537783 | ||

BFAR
Genomic Map