siRNA Details
Gene Symbol: RXFP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016568.3 | NP_057652.1 | 1 | 606 | ||
GenBank mRNA | AY394501.1 | - | 1 | 251 | ||
BC095526.1 | - | 1 | 325 | 30706560 | ||
BC113438.1 | - | 1 | 327 | 8322490 | ||
BC113440.1 | - | 1 | 327 | 8322492 | ||
D88437.1 | - | 1 | 611 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
