siRNA Details
| Gene Symbol: PCDH12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016580.3 | NP_057664.1 | 1 | 1901 | ||
| GenBank mRNA | AB026893.1 | - | 1 | 747 | ||
| AF231025.1 | - | 1 | 996 | |||
| AF240635.1 | - | 1 | 1901 | |||
| AK024140.1 | - | 1 | 1039 | |||
| AK027282.1 | - | 1 | 992 | |||
| AK226049.1 | - | 1 | 998 | |||
| AK315029.1 | - | 1 | 998 | |||
| AY358428.1 | - | 1 | 1018 | |||
| BC042634.1 | - | 1 | 951 | 5177606 | ||
| BC052973.1 | - | 1 | 1008 | 5174442 | ||

PCDH12
Genomic Map