siRNA Details
| Gene Symbol: NRN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278710.1 | NP_001265639.1 | 4 | 564 | ||
| NM_001278711.1 | NP_001265640.1 | 4 | 803 | |||
| NM_016588.2 | NP_057672.1 | 3 | 1062 | |||
| XM_006715106.3 | XP_006715169.1 | 5 | 1453 | |||
| XM_006715107.3 | XP_006715170.1 | 3 | 1243 | |||
| GenBank mRNA | AF114833.1 | - | 3 | 593 | ||
| AF136631.1 | - | 3 | 579 | |||
| AJ420483.1 | - | 3 | 536 | |||
| AK093824.1 | - | 3 | 1257 | |||
| AK314095.1 | - | 3 | 615 | |||
| AL541038.3 | - | 3 | 647 | |||
| BC002683.2 | - | 3 | 540 | 3605775 | ||
| BC042019.1 | - | 3 | 1063 | 5312094 | ||
| BF970432.1 | - | 3 | 455 | |||
| BX340783.2 | - | 4 | 799 | |||

NRN1
Genomic Map