siRNA Details
| Gene Symbol: GCNT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016591.2 | NP_057675.1 | 1 | 1437 | ||
| GenBank mRNA | AF132035.1 | - | 1 | 1436 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GCNT4| Gene Symbol: GCNT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016591.2 | NP_057675.1 | 1 | 1437 | ||
| GenBank mRNA | AF132035.1 | - | 1 | 1436 | ||