siRNA Details
| Gene Symbol: FAM13B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101800.2 | NP_001095270.1 | 15 | 2296 | ||
| NM_001101801.2 | NP_001095271.1 | 15 | 2170 | |||
| NM_016603.3 | NP_057687.2 | 15 | 2296 | |||
| XM_005272007.1 | XP_005272064.1 | 16 | 2362 | |||
| XM_006714647.1 | XP_006714710.1 | 16 | 2362 | |||
| XM_006714648.1 | XP_006714711.1 | 16 | 2114 | |||
| XM_006714649.1 | XP_006714712.1 | 15 | 2170 | |||
| XM_011543448.1 | XP_011541750.1 | 17 | 2155 | |||
| XM_011543449.1 | XP_011541751.1 | 17 | 2459 | |||
| XM_011543450.1 | XP_011541752.1 | 16 | 2071 | |||
| XM_017009549.1 | XP_016865038.1 | 15 | 2321 | |||
| XM_017009550.1 | XP_016865039.1 | 15 | 2005 | |||
| XM_017009551.1 | XP_016865040.1 | 14 | 2104 | |||
| GenBank mRNA | AF251038.1 | - | 15 | 2215 | ||
| AK314224.1 | - | 15 | 2272 | |||
| AL833699.1 | - | 2 | 320 | |||
| BC041335.1 | - | 15 | 2148 | |||
| BC048965.1 | - | 15 | 2268 | 5265611 | ||
| BC056887.1 | - | 14 | 1871 | 6148274 | ||

FAM13B
Genomic Map