siRNA Details
| Gene Symbol: ARMCX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016608.1 | NP_057692.1 | 4 | 1319 | ||
| GenBank mRNA | AB039670.1 | - | 4 | 1319 | ||
| AF248963.1 | - | 4 | 1252 | |||
| AK074785.1 | - | 4 | 1269 | |||
| AK222578.1 | - | 4 | 1296 | |||
| BC002691.2 | - | 4 | 1258 | 3605690 | ||
| CR749609.1 | - | 5 | 969 | |||

ARMCX1
Genomic Map