siRNA Details
| Gene Symbol: KLRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291822.1 | NP_001278751.1 | 4 | 476 | ||
| NM_016523.2 | NP_057607.1 | 5 | 626 | |||
| NR_120305.1 | 3 | 336 | ||||
| GenBank mRNA | AF175206.1 | - | 5 | 626 | ||
| AF175207.1 | - | 4 | 476 | |||
| AF267244.1 | - | 3 | 336 | |||
| AJ305370.1 | - | 5 | 609 | |||
| BC098166.1 | - | 4 | 465 | 40015535 | ||
| BC098257.1 | - | 4 | 446 | 40015534 | ||
| BC098354.1 | - | 5 | 586 | 40015531 | ||

KLRF1
Genomic Map