siRNA Details
| Gene Symbol: KRT76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015848.4 | NP_056932.2 | 7 | 1389 | ||
| GenBank mRNA | AK299390.1 | - | 7 | 1389 | ||
| M99063.1 | - | 7 | 1366 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT76| Gene Symbol: KRT76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015848.4 | NP_056932.2 | 7 | 1389 | ||
| GenBank mRNA | AK299390.1 | - | 7 | 1389 | ||
| M99063.1 | - | 7 | 1366 | |||