siRNA Details
Gene Symbol: DCDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195610.1 | NP_001182539.1 | 7, 8 | 959 | ||
NM_016356.4 | NP_057440.2 | 6, 7 | 1052 | |||
GenBank mRNA | AB032980.2 | - | 6, 7 | 750 | ||
AF181721.1 | - | 6, 7 | 1052 | |||
AK027036.1 | - | 6, 7 | 868 | |||
BC050704.1 | - | 7, 8 | 959 | 5758845 |
