siRNA Details
| Gene Symbol: ANKFY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001257999.1 | NP_001244928.1 | 6 | 900 | ||
| NM_016376.3 | NP_057460.3 | 6 | 695 | |||
| NR_047571.1 | 5 | 576 | ||||
| XM_005256679.3 | XP_005256736.1 | 6 | 742 | |||
| XM_011523926.1 | XP_011522228.1 | 6 | 798 | |||
| XM_011523928.2 | XP_011522230.1 | 6 | 780 | |||
| XM_011523933.1 | XP_011522235.1 | 2 | 184 | |||
| XM_017024733.1 | XP_016880222.1 | 6 | 995 | |||
| GenBank mRNA | AB033081.2 | - | 6 | 900 | ||
| AB037360.1 | - | 6 | 663 | |||
| AK025483.1 | - | 6 | 672 | |||
| AK057047.1 | - | 5 | 553 | |||
| AK225170.1 | - | 6 | 672 | |||
| AK292930.1 | - | 6 | 672 | |||
| AL706226.1 | - | 6 | 695 | |||
| BC052308.1 | - | 6 | 668 | |||

ANKFY1
Genomic Map