siRNA Details
| Gene Symbol: SPOUT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016390.3 | NP_057474.2 | 7 | 661 | ||
| XM_017014804.1 | XP_016870293.1 | 8 | 928 | |||
| GenBank mRNA | AF161459.1 | - | 7 | 632 | ||
| AL110193.1 | - | 7 | 661 | |||
| BC010579.1 | - | 7 | 623 | 4213299 | ||
| BC021273.1 | - | 7 | 634 | 4843384 | ||
| BC033677.1 | - | 7 | 629 | 5176852 | ||
| BC039590.1 | - | 7 | 646 | 5757547 | ||
| BC046133.1 | - | 7 | 626 | 5725479 | ||
| BC063644.1 | - | 7 | 629 | 4539701 | ||

SPOUT1
Genomic Map