siRNA Details
| Gene Symbol: SPOUT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016390.3 | NP_057474.2 | 10, 11 | 951 | ||
| XM_017014804.1 | XP_016870293.1 | 11, 12 | 1218 | |||
| GenBank mRNA | AF161459.1 | - | 10, 11 | 921 | ||
| AL110193.1 | - | 10, 11 | 951 | |||
| BC010579.1 | - | 10, 11 | 913 | 4213299 | ||
| BC021273.1 | - | 10, 11 | 924 | 4843384 | ||
| BC033677.1 | - | 10, 11 | 919 | 5176852 | ||
| BC039590.1 | - | 10, 11 | 936 | 5757547 | ||
| BC046133.1 | - | 10, 11 | 916 | 5725479 | ||
| BC063644.1 | - | 10, 11 | 919 | 4539701 | ||

SPOUT1
Genomic Map