siRNA Details
| Gene Symbol: RTCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014306.4 | NP_055121.1 | 11 | 1519 | ||
| GenBank mRNA | AF155658.1 | - | 11 | 1467 | ||
| AF161466.1 | - | 11 | 1478 | |||
| AK297908.1 | - | 677 | ||||
| AK312503.1 | - | 11 | 1479 | |||
| AL050255.1 | - | 11 | 1463 | |||
| AL137272.1 | - | 4 | 465 | |||
| BC000151.1 | - | 11 | 1457 | 2984824 | ||
| BC002970.1 | - | 11 | 1457 | 3533883 | ||
| BC010308.1 | - | 11 | 1469 | 3451576 | ||
| BC016707.1 | - | 11 | 1484 | 4102148 | ||
| BU615556.1 | - | 2 | 562 | |||
| CR456450.1 | - | 11 | 1434 | |||
| CR457175.1 | - | 11 | 1388 | |||

RTCB
Genomic Map