siRNA Details
| Gene Symbol: UFC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016406.3 | NP_057490.2 | 5 | 611 | ||
| XM_017001450.1 | XP_016856939.1 | 6 | 456 | |||
| XM_017001451.1 | XP_016856940.1 | 6 | 720 | |||
| GenBank mRNA | AB154405.1 | - | 5 | 357 | ||
| AF151884.1 | - | 5 | 578 | |||
| AF161504.1 | - | 5 | 597 | |||
| AK292776.1 | - | 5 | 563 | |||
| BC005187.1 | - | 5 | 603 | 3961569 | ||
| CF128121.1 | - | 5 | 610 | |||

UFC1
Genomic Map