siRNA Details
| Gene Symbol: GSKIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271904.1 | NP_001258833.1 | 3 | 744 | ||
| NM_001271905.1 | NP_001258834.1 | 3 | 297 | |||
| NM_001271906.1 | NP_001258835.1 | 2 | 229 | |||
| NM_016472.4 | NP_057556.2 | 3 | 330 | |||
| XM_017021365.1 | XP_016876854.1 | 3 | 438 | |||
| GenBank mRNA | AF151044.1 | - | 3 | 248 | ||
| AK000796.1 | - | 3 | 256 | |||
| AK094654.1 | - | 3 | 746 | |||
| AK314423.1 | - | 3 | 256 | |||
| BC004818.2 | - | 2 | 216 | 3449501 | ||
| DA488110.1 | - | 3 | 267 | |||
| HY051680.1 | - | 3 | 331 | |||
| HY108584.1 | - | 2 | 205 | |||

GSKIP
Genomic Map