siRNA Details
| Gene Symbol: ESF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276380.1 | NP_001263309.1 | 8 | 1715 | ||
| NM_016649.3 | NP_057733.2 | 8 | 1726 | |||
| XM_017027874.1 | XP_016883363.1 | 8 | 3178 | |||
| GenBank mRNA | AF068285.1 | - | 7 | 1083 | ||
| AK000375.1 | - | 8 | 1727 | |||
| AK022369.1 | - | 8 | 1677 | |||
| AK074299.1 | - | 7 | 1212 | |||
| BC020952.1 | - | 7 | 1345 | |||
| BC030513.1 | - | 7 | 1676 | |||
| BC046107.1 | - | 7 | 1665 | |||

ESF1
Genomic Map