siRNA Details
| Gene Symbol: CUTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014433.2 | NP_001014433.1 | 3 | 457 | ||
| NM_001014837.1 | NP_001014837.1 | 3 | 706 | |||
| NM_001014838.1 | NP_001014838.1 | 3 | 675 | |||
| NM_001014840.1 | NP_001014840.1 | 3 | 585 | |||
| NM_015921.2 | NP_057005.1 | 2 | 848 | |||
| GenBank mRNA | AF106943.1 | - | 2 | 485 | ||
| AF230924.1 | - | 2 | 271 | |||
| AK311556.1 | - | 1 | 837 | |||
| AL533788.3 | - | 2 | 668 | |||
| BC005890.2 | - | 3 | 282 | 3505574 | ||
| BC107751.1 | - | 2 | 485 | 6714433 | ||
| BU540844.1 | - | 463 | ||||
| CB106834.1 | - | 3 | 411 | |||
| CN363642.1 | - | 3 | 487 | |||

CUTA
Genomic Map