siRNA Details
| Gene Symbol: LIPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204830.1 | NP_001191759.1 | 3 | 989 | ||
| NM_015929.3 | NP_057013.1 | 3 | 791 | |||
| NM_145197.2 | NP_660198.1 | 4 | 919 | |||
| NM_145198.2 | NP_660199.1 | 4 | 907 | |||
| NM_145199.2 | NP_660200.1 | 2 | 665 | |||
| NR_037935.1 | 4 | 1052 | ||||
| NR_037936.1 | 3 | 728 | ||||
| GenBank mRNA | BC009772.1 | - | 3 | 732 | 4091817 | |
| BC070145.1 | - | 3 | 769 | 4671058 | ||

LIPT1
Genomic Map