siRNA Details
| Gene Symbol: MRPL48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318498.1 | NP_001305427.1 | 9 | 753 | ||
| NM_001318499.1 | NP_001305428.1 | 9 | 818 | |||
| NM_001318500.1 | NP_001305429.1 | 6 | 422 | |||
| NM_016055.5 | NP_057139.1 | 8 | 695 | |||
| NR_134658.1 | 7 | 657 | ||||
| NR_134659.1 | 10 | 948 | ||||
| NR_134660.1 | 8 | 813 | ||||
| NR_134661.1 | 6 | 503 | ||||
| XM_017017919.1 | XP_016873408.1 | 9 | 788 | |||
| GenBank mRNA | AF151876.1 | - | 8 | 1251 | ||
| AF161408.1 | - | 8 | 698 | |||
| AK297746.1 | - | 9 | 753 | |||
| AK310005.1 | - | 10 | 927 | |||
| BC009764.1 | - | 8 | 688 | 4093230 | ||
| BI549412.1 | - | 7 | 650 | |||
| BQ446725.1 | - | 1 | 299 | |||
| BU600573.1 | - | 6 | 386 | |||
| BX354271.2 | - | 8 | 778 | |||
| CF529680.1 | - | 1 | 299 | |||
| CF541319.1 | - | 1 | 293 | |||

MRPL48
Genomic Map