siRNA Details
| Gene Symbol: SARAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284239.1 | NP_001271168.1 | 4 | 1044 | ||
| NM_016127.5 | NP_057211.4 | 5 | 1223 | |||
| XM_017013567.1 | XP_016869056.1 | 5 | 1473 | |||
| XM_017013568.1 | XP_016869057.1 | 5 | 901 | |||
| XM_017013569.1 | XP_016869058.1 | 5 | 901 | |||
| XM_017013570.1 | XP_016869059.1 | 4 | 983 | |||
| GenBank mRNA | AB028926.1 | - | 5 | 1083 | ||
| AF078855.1 | - | 5 | 1051 | |||
| AF100748.1 | - | 5 | 980 | |||
| AF490252.1 | - | 5 | 964 | |||
| AK025927.1 | - | 5 | 1133 | |||
| AK075340.1 | - | 5 | 1081 | |||
| AK130057.1 | - | 4 | 831 | |||
| AK222632.1 | - | 5 | 980 | |||
| AK295360.1 | - | 6 | 1028 | |||
| AK314659.1 | - | 5 | 1105 | |||
| AK315956.1 | - | 4 | 801 | |||
| AY359104.1 | - | 5 | 1148 | |||
| AY454545.1 | - | 4 | 800 | |||
| BC015012.1 | - | 5 | 1052 | 3908758 | ||
| JQ348891.1 | - | 5 | 1092 | |||
| JQ348892.1 | - | 4 | 784 | |||

SARAF
Genomic Map