siRNA Details
| Gene Symbol: TRAPPC2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318524.1 | NP_001305453.1 | 4 | 389 | ||
| NM_001318525.1 | NP_001305454.1 | 4 | 389 | |||
| NM_001318526.1 | NP_001305455.1 | 4 | 454 | |||
| NM_001318527.1 | NP_001305456.1 | 4 | 705 | |||
| NM_001318528.1 | NP_001305457.1 | 4 | 366 | |||
| NM_001318529.1 | NP_001305458.1 | 4 | 705 | |||
| NM_001318530.1 | NP_001305459.1 | 4 | 366 | |||
| NM_001318532.1 | NP_001305461.1 | 4 | 705 | |||
| NM_016209.4 | NP_057293.1 | 4 | 389 | |||
| NR_134670.1 | 5 | 576 | ||||
| NR_134671.1 | 5 | 505 | ||||
| XM_017023270.1 | XP_016878759.1 | 4 | 436 | |||
| GenBank mRNA | AF089106.1 | - | 4 | 382 | ||
| AF161524.1 | - | 4 | 344 | |||
| AK126779.1 | - | 4 | 348 | |||
| AK307253.1 | - | 5 | 505 | |||
| AK310805.1 | - | 4 | 660 | |||
| AK311885.1 | - | 4 | 358 | |||
| AX771012.1 | - | 4 | 382 | |||
| AX771013.1 | - | 4 | 344 | |||
| AX771014.1 | - | 3 | 250 | |||
| BC011369.1 | - | 4 | 336 | 3868798 | ||
| BC018024.1 | - | 4 | 343 | 4287920 | ||
| BC105809.1 | - | 4 | 355 | 6745065 | ||
| BI754306.1 | - | 5 | 518 | |||
| BX419849.2 | - | 4 | 334 | |||
| DA209193.1 | - | 4 | 366 | |||
| DA293122.1 | - | 4 | 389 | |||
| DA833474.1 | - | 4 | 328 | |||

TRAPPC2L
Genomic Map