siRNA Details
| Gene Symbol: CSNK1G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022048.3 | NP_071331.2 | 2 | 514 | ||
| GenBank mRNA | AB042562.1 | - | 1 | 162 | ||
| AB042563.1 | - | 1 | 146 | |||
| AF223357.2 | - | 1 | 232 | |||
| AK225960.1 | - | 2 | 444 | |||
| AL832551.1 | - | 2 | 292 | |||
| BC130613.1 | - | 2 | 287 | 40146644 | ||
| BC136504.1 | - | 2 | 287 | 9020493 | ||
| CF593592.1 | - | 2 | 472 | |||
| DB094870.1 | - | 2 | 514 | |||

CSNK1G1
Genomic Map