siRNA Details
| Gene Symbol: SNTG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018968.3 | NP_061841.2 | 13 | 1179 | ||
| XM_011510366.2 | XP_011508668.1 | 11 | 1216 | |||
| XM_017004361.1 | XP_016859850.1 | 13 | 1141 | |||
| XM_017004362.1 | XP_016859851.1 | 13 | 1147 | |||
| XM_017004363.1 | XP_016859852.1 | 13 | 1328 | |||
| XM_017004364.1 | XP_016859853.1 | 10 | 4469 | |||
| XM_017004365.1 | XP_016859854.1 | 13 | 1142 | |||
| XM_017004366.1 | XP_016859855.1 | 13 | 1142 | |||
| XM_017004367.1 | XP_016859856.1 | 14 | 1217 | |||
| XM_017004373.1 | XP_016859862.1 | 8 | 1783 | |||
| GenBank mRNA | AJ003029.1 | - | 13 | 1181 | ||
| BC125251.1 | - | 7 | 727 | 40025232 | ||

SNTG2
Genomic Map