siRNA Details
| Gene Symbol: SNTG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018968.3 | NP_061841.2 | 14 | 1367 | ||
| XM_011510366.2 | XP_011508668.1 | 12 | 1404 | |||
| XM_017004361.1 | XP_016859850.1 | 14 | 1329 | |||
| XM_017004362.1 | XP_016859851.1 | 14 | 1335 | |||
| XM_017004363.1 | XP_016859852.1 | 14 | 1516 | |||
| XM_017004364.1 | XP_016859853.1 | 11 | 4657 | |||
| XM_017004365.1 | XP_016859854.1 | 14 | 1330 | |||
| XM_017004366.1 | XP_016859855.1 | 14 | 1330 | |||
| XM_017004367.1 | XP_016859856.1 | 15 | 1405 | |||
| XM_017004373.1 | XP_016859862.1 | 9 | 1971 | |||
| GenBank mRNA | AJ003029.1 | - | 14 | 1369 | ||
| BC125251.1 | - | 8 | 915 | 40025232 | ||

SNTG2
Genomic Map