siRNA Details
| Gene Symbol: CYTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018659.2 | NP_061129.1 | 3 | 329 | ||
| XM_017008299.1 | XP_016863788.1 | 4 | 444 | |||
| GenBank mRNA | AF193766.1 | - | 3 | 332 | ||
| AF274956.1 | - | 3 | 233 | |||
| AY359101.1 | - | 3 | 330 | |||
| BC031391.1 | - | 3 | 305 | 5186334 | ||
| T83161.1 | - | 1 | 61 | |||

CYTL1
Genomic Map