siRNA Details
| Gene Symbol: CYTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018659.2 | NP_061129.1 | 4 | 377 | ||
| XM_017008299.1 | XP_016863788.1 | 5 | 492 | |||
| GenBank mRNA | AF193766.1 | - | 4 | 380 | ||
| AY359101.1 | - | 4 | 378 | |||
| BC031391.1 | - | 4 | 353 | 5186334 | ||
| T83161.1 | - | 2 | 109 | |||

CYTL1
Genomic Map