siRNA Details
| Gene Symbol: RIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242581.1 | NP_001229510.1 | 8 | 1021 | ||
| NM_018993.3 | NP_061866.1 | 7 | 761 | |||
| XM_005260731.2 | XP_005260788.1 | 9 | 1217 | |||
| XM_005260733.2 | XP_005260790.1 | 5 | 569 | |||
| XM_006723574.3 | XP_006723637.1 | 10 | 1199 | |||
| XM_006723575.3 | XP_006723638.1 | 9 | 985 | |||
| XM_006723577.2 | XP_006723640.1 | 8 | 834 | |||
| XM_011529255.2 | XP_011527557.2 | 8 | 1294 | |||
| XM_011529257.1 | XP_011527559.1 | 8 | 890 | |||
| XM_011529258.2 | XP_011527560.1 | 8 | 902 | |||
| XM_011529259.2 | XP_011527561.1 | 7 | 814 | |||
| XM_017027887.1 | XP_016883376.1 | 11 | 1540 | |||
| XM_017027888.1 | XP_016883377.1 | 12 | 1612 | |||
| XM_017027889.1 | XP_016883378.1 | 7 | 1141 | |||
| XM_017027890.1 | XP_016883379.1 | 9 | 972 | |||
| XM_017027891.1 | XP_016883380.1 | 9 | 1010 | |||
| XM_017027892.1 | XP_016883381.1 | 7 | 825 | |||
| XM_017027893.1 | XP_016883382.1 | 8 | 1319 | |||
| GenBank mRNA | AB060339.1 | - | 7 | 725 | ||
| AK094884.1 | - | 8 | 1022 | |||
| AL136924.1 | - | 6 | 642 | |||

RIN2
Genomic Map