siRNA Details
| Gene Symbol: TRMT13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019083.2 | NP_061956.2 | 10 | 1055 | ||
| NR_135078.1 | 10 | 814 | ||||
| XM_005270945.3 | XP_005271002.1 | 10 | 1083 | |||
| XM_005270946.2 | XP_005271003.1 | 9 | 975 | |||
| XM_017001504.1 | XP_016856993.1 | 10 | 2409 | |||
| XM_017001505.1 | XP_016856994.1 | 10 | 2398 | |||
| GenBank mRNA | AK002081.1 | - | 10 | 1042 | ||
| AK298941.1 | - | 10 | 799 | |||
| BC075811.1 | - | 10 | 1052 | 5104751 | ||

TRMT13
Genomic Map