siRNA Details
| Gene Symbol: FAM105A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019018.2 | NP_061891.1 | 2, 3 | 330 | ||
| XM_011514056.2 | XP_011512358.1 | 2, 3 | 312 | |||
| GenBank mRNA | AK001989.1 | - | 2, 3 | 289 | ||
| AK222731.1 | - | 2, 3 | 317 | |||
| BC011524.1 | - | 2, 3 | 256 | 3872837 | ||
| BX342178.2 | - | 2, 3 | 337 | |||

FAM105A
Genomic Map