siRNA Details
| Gene Symbol: HEATR5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019024.2 | NP_061897.1 | 9 | 1417 | ||
| XM_006712034.2 | XP_006712097.1 | 9 | 1356 | |||
| XM_006712035.3 | XP_006712098.1 | 9 | 1420 | |||
| XM_011532934.2 | XP_011531236.1 | 9 | 1884 | |||
| XM_017004378.1 | XP_016859867.1 | 9 | 1356 | |||
| XM_017004379.1 | XP_016859868.1 | 10 | 1794 | |||
| XM_017004380.1 | XP_016859869.1 | 10 | 1567 | |||
| GenBank mRNA | AK096637.1 | - | 9 | 1356 | ||
| BC150605.1 | - | 9 | 1402 | 9056975 | ||
| BX538008.1 | - | 3 | 536 | |||

HEATR5B
Genomic Map