siRNA Details
| Gene Symbol: PUS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318163.1 | NP_001305092.1 | 1 | 366 | ||
| NM_001318164.1 | NP_001305093.1 | 2 | 573 | |||
| NM_019042.4 | NP_061915.2 | 2 | 597 | |||
| XM_005250462.3 | XP_005250519.1 | 2 | 605 | |||
| XM_011516336.2 | XP_011514638.1 | 2 | 600 | |||
| XM_017012367.1 | XP_016867856.1 | 2 | 533 | |||
| XM_017012368.1 | XP_016867857.1 | 2 | 609 | |||
| GenBank mRNA | AB067484.1 | - | 1 | 293 | ||
| AK091283.1 | - | 2 | 577 | |||
| AK128629.1 | - | 2 | 559 | |||
| BC011396.1 | - | 2 | 553 | 3870711 | ||
| CN481800.1 | - | 2 | 573 | |||

PUS7
Genomic Map