siRNA Details
| Gene Symbol: CCDC93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019044.4 | NP_061917.3 | 15, 16 | 1401 | ||
| XM_011511359.1 | XP_011509661.1 | 15, 16 | 1398 | |||
| XM_011511361.1 | XP_011509663.1 | 14, 15 | 1155 | |||
| XM_011511362.1 | XP_011509664.1 | 14, 15 | 1269 | |||
| GenBank mRNA | AB209993.1 | - | 15, 16 | 1320 | ||
| AK001858.1 | - | 3, 4 | 195 | |||
| AK025611.1 | - | 14, 15 | 1169 | |||
| AK057926.1 | - | 15, 16 | 1262 | |||
| AK290722.1 | - | 15, 16 | 1308 | |||
| AK309040.1 | - | 15, 16 | 1308 | |||
| BC028609.1 | - | 15, 16 | 1374 | 4826022 | ||
| BC093018.1 | - | 15, 16 | 1291 | 30337330 | ||

CCDC93
Genomic Map