siRNA Details
| Gene Symbol: ASNSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019048.2 | NP_061921.1 | 5 | 2022 | ||
| XM_017004381.1 | XP_016859870.1 | 4 | 1753 | |||
| XM_017004382.1 | XP_016859871.1 | 4 | 2538 | |||
| GenBank mRNA | AB075481.1 | - | 2 | 1471 | ||
| AK000759.1 | - | 5 | 2001 | |||
| AY116969.1 | - | 2 | 1588 | |||
| BC001243.1 | - | 5 | 1927 | 3453136 | ||

ASNSD1
Genomic Map