siRNA Details
| Gene Symbol: MIER2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017550.1 | NP_060020.1 | 5, 6 | 495 | ||
| XM_005259582.4 | XP_005259639.1 | 5, 6 | 1079 | |||
| XM_006722769.3 | XP_006722832.1 | 5, 6 | 1083 | |||
| XM_011528072.2 | XP_011526374.1 | 5, 6 | 1081 | |||
| XM_011528073.1 | XP_011526375.1 | 5, 6 | 497 | |||
| XM_011528074.2 | XP_011526376.1 | 5, 6 | 1085 | |||
| XM_011528075.2 | XP_011526377.1 | 5, 6 | 1602 | |||
| XM_011528077.2 | XP_011526379.1 | 6, 7 | 3443 | |||
| XM_011528081.2 | XP_011526383.1 | 3, 4 | 339 | |||
| XM_011528082.2 | XP_011526384.1 | 3, 4 | 270 | |||
| GenBank mRNA | AB033019.1 | - | 483 | |||
| BC028203.1 | - | 5, 6 | 495 | 5201526 | ||

MIER2
Genomic Map