siRNA Details
| Gene Symbol: RC3H2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100588.1 | NP_001094058.1 | 18 | 3312 | ||
| NM_018835.2 | NP_061323.2 | 18 | 3312 | |||
| GenBank mRNA | AF255303.1 | - | 17 | 3672 | ||
| AF255304.1 | - | 6 | 756 | |||
| AK000308.1 | - | 16 | 2746 | |||
| AK000720.1 | - | 5 | 539 | |||
| AK027042.1 | - | 9 | 1549 | |||
| AK226010.1 | - | 16 | 2746 | |||
| AL833177.1 | - | 19 | 3169 | |||
| AL834431.1 | - | 12 | 2063 | |||
| AW291376.1 | - | 1 | 266 | |||

RC3H2
Genomic Map